Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-dimethylphenyl)methanol 97%

Supplier: AMBEED, INC
Warning

A344600-1G A344600-10G A344600-250MG A344600-25G A344600-5G A344600-1G
103825-910EA 10 USD InStock
103825-910 103836-890 76694-026 76694-028 76694-030 76839-942 77306-564
(2,6-dimethylphenyl)methanol 97%
(2,6-dimethylphenyl)methanol
Precaución: for Research Use Only


Formula: C₉H₁₂O
MW: 136.194 g/mol
Boiling Pt: 225.9 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00128004
CAS Number: 62285-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty