Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2'-Amino-[1,1'-biphenyl]-2-yl)(dicyclohexyl(2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl)phosphoranyl)palladium(III) chloride 98%

Supplier: AMBEED, INC
Warning

A127208-1G A127208-10G A127208-250MG A127208-1G
103800-932EA 26 USD InStock
103800-932 77009-722 77009-724 77009-726 77313-178
(2'-Amino-[1,1'-biphenyl]-2-yl)(dicyclohexyl(2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl)phosphoranyl)palladium(III) chloride 98%
(2'-Amino-[1,1'-biphenyl]-2-yl)(dicyclohexyl(2',6'-diisopropoxy-[1,1'-biphenyl]-2-yl)phosphoranyl)palladium(III) chloride
Precaución: for Research Use Only


Formula: C₄₂H₅₄ClNO₂PPd
Storage Temperature: Refrigerator
MDL Number: MFCD20264899
CAS Number: 1375325-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty