Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A277117-1G A277117-5G A277117-1G A277117-5G
103813-334EA 45 USD InStock
103813-334 103853-270 77093-146 77093-148
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride 97%
2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀BNO₄·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD02258941
CAS Number: 380430-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty