Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Amino-5-chlorophenyl)(2,6-difluorophenyl)methanone 98%

Supplier: AMBEED, INC
Warning

A101793-1G A101793-5G A101793-10G A101793-250MG A101793-25G A101793-1G A101793-5G
103796-140EA 27 USD InStock
103796-140 103821-828 77093-530 77093-532 77093-534 77282-834 77282-836
(2-Amino-5-chlorophenyl)(2,6-difluorophenyl)methanone 98%
(2-Amino-5-chlorophenyl)(2,6-difluorophenyl)methanone
Precaución: for Research Use Only


Formula: C₁₃H₈ClF₂NO
MW: 267.6600 g/mol
Boiling Pt: 435.2 °C (760 mm Hg)
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD06659048
CAS Number: 28910-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty