Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Amino-5-methoxyphenyl)methanol 97%

Supplier: AMBEED, INC
Warning

A152154-1G A152154-5G A152154-250MG A152154-1G
103804-352EA 34 USD InStock
103804-352 103842-510 76871-642 77299-928
(2-Amino-5-methoxyphenyl)methanol 97%
(2-Amino-5-methoxyphenyl)methanol
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂
Boiling Pt: 328.1±27.0 °C (760 mm Hg)
Melting Pt: 88…90 °C
Flash Pt: 152.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09966091
CAS Number: 55414-72-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty