Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Amino-6-chlorophenyl)methanol 97%

Supplier: AMBEED, INC
Warning

A141345-1G A141345-100G A141345-10G A141345-250MG A141345-1G A141345-25G A141345-5G
103802-978EA 9 USD InStock
103802-978 77137-328 77137-330 77137-332 77316-126 77316-128 77316-130
(2-Amino-6-chlorophenyl)methanol 97%
(2-Amino-6-chlorophenyl)methanol
Precaución: for Research Use Only


Formula: C₇H₈ClNO
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD09029932
CAS Number: 39885-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty