Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Amino-1,3-thiazol-5-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A253025-1G A253025-100MG A253025-250MG
103812-070EA 53 USD InStock
103812-070 103851-918 76976-718 77881-683
(2-Amino-1,3-thiazol-5-yl)methanol 97%
(2-Amino-1,3-thiazol-5-yl)methanol
Precaución: for Research Use Only


Formula: C₄H₆N₂OS
Boiling Pt: 340.738 °C (760 mm Hg)
Melting Pt: 102…103 °C
Flash Pt: 159.873 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203018
CAS Number: 131184-73-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty