Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromoethoxy)-tert-butyldimethylsilane 96%

Supplier: AMBEED, INC
Warning

A206034-1G A206034-5G A206034-25G A206034-5G A206034-100G A206034-10G A206034-500G A206034-1G
103817-682EA 5 USD InStock
103817-682 103848-356 77086-592 77086-594 77139-500 77139-502 77139-504 77302-980
(2-Bromoethoxy)-tert-butyldimethylsilane 96%
(2-Bromoethoxy)-tert-butyldimethylsilane
Precaución: for Research Use Only


Formula: C₈H₁₉BrOSi
Boiling Pt: 191.2 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209550
CAS Number: 86864-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty