Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A159659-1G A159659-5G A159659-100G A159659-10G A159659-1G A159659-25G A159659-5G
103805-274EA 5 USD InStock
103805-274 103843-496 77273-260 77273-262 77319-388 77319-390 77319-392
2-Bromophenylboronic acid 98%
2-Bromophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 182…186 °C
Flash Pt: 152.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01114672
CAS Number: 244205-40-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty