Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A408352-1G A408352-25G A408352-5G A408352-100G A408352-10G
103827-184EA 5 USD InStock
103827-184 103838-230 76841-740 76841-742 76841-744 77115-794 77115-796
2-Chlorophenylboronic acid 97%
2-Chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BClO₂
MW: 156.38
Boiling Pt: 306.3 °C (760 mm Hg)
Melting Pt: 92…102 °C
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00674012
CAS Number: 3900-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty