Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloropyrimidin-5-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A215465-1G A215465-5G A215465-100MG A215465-250MG A215465-1G A215465-25G A215465-5G A215465-10G
103818-490EA 25 USD InStock
103818-490 103849-196 76814-712 76814-714 77087-722 77087-724 77087-726 77671-928
(2-Chloropyrimidin-5-yl)methanol 97%
(2-Chloropyrimidin-5-yl)methanol
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
Boiling Pt: 341.8±17.0 °C (760 mm Hg)
Melting Pt: 112…116 °C
Flash Pt: 160.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16659172
CAS Number: 1046816-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty