Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A228147-1G A228147-1G A228147-5G A228147-100G A228147-100MG A228147-10G A228147-250MG
103819-366EA 8 USD InStock
103819-366 103850-274 77089-068 77089-070 77139-988 77139-990 77139-992 77139-994
2-Cyanophenylboronic acid 97%
2-Cyanophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BNO₂
MW: 146.94
Melting Pt: 126…127 °C
Flash Pt: 181.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01632208
CAS Number: 138642-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty