Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methylphenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A350542-1G A350542-5G A350542-100MG A350542-5G
103826-038EA 45 USD InStock
103826-038 103837-032 76876-290 77306-610
2-Fluoro-6-methylphenylboronic acid 95%
2-Fluoro-6-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₂
MW: 153.947 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13194360
CAS Number: 887471-69-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty