Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A292057-5G A292057-25G A292057-100G A292057-10G A292057-1G A292057-500G A292057-5G
103835-064EA 7 USD InStock
103835-064 76837-660 76875-484 76875-486 76875-488 76875-490 77305-614
2-Fluorophenylboronic acid 98%
2-Fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Boiling Pt: 267.8 °C (760 mm Hg)
Melting Pt: 101…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00674013
CAS Number: 1993-03-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty