Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorophenylhydrazinium chloride 98%

Supplier: AMBEED, INC
Warning

A314689-5G A314689-25G A314689-10G A314689-100G
103835-966EA 6 USD InStock
103835-966 76838-786 77290-196 77748-614
2-Fluorophenylhydrazinium chloride 98%
2-Fluorophenylhydrazinium chloride
Precaución: for Research Use Only


Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 200…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00012927
CAS Number: 2924-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty