Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-pyridineboronic acid 98%

Supplier: AMBEED, INC
Warning

A105899-1G A105899-5G A105899-100G A105899-10G A105899-1G A105899-5G
103796-962EA 9 USD InStock
103796-962 103822-708 77008-596 77008-598 77283-408 77283-410
2-Fluoro-3-pyridineboronic acid 98%
2-Fluoro-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₅BFNO₂
Boiling Pt: 322.6 °C (760 mm Hg)
Flash Pt: 148.9 °C
Storage Temperature: Freezer
MDL Number: MFCD03411557
CAS Number: 174669-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty