Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A162326-1G A162326-5G A162326-100G A162326-10G A162326-500G A162326-25G A162326-1G A162326-5G
103805-604EA 8 USD InStock
103805-604 103843-854 77110-742 77110-744 77110-746 77300-588 77319-888 77319-890
2-Nitrophenylboronic acid 97%
2-Nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Boiling Pt: 374.7 °C (760 mm Hg)
Melting Pt: 155…160 °C
Flash Pt: 180.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00161358
CAS Number: 5570-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty