Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chloropurine riboside 95%

Supplier: AMBEED, INC
Warning

A266115-1G A266115-5G A266115-100G A266115-1G A266115-25G
103812-782EA 6 USD InStock
103812-782 103852-682 76772-010 77092-380 77092-382
2-Amino-6-chloropurine riboside 95%
2-Amino-6-chloropurine riboside
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClN₅O₄
MW: 301.69 g/mol
Boiling Pt: 729.9 °C (760 mm Hg)
Melting Pt: 165…167 °C
Storage Temperature: Freezer
MDL Number: MFCD00005735
CAS Number: 2004-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty