Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3R,4S,5R)-6-Oxohexane-1,2,3,4,5-pentayl pentaacetate 97%

Supplier: AMBEED, INC
Warning

A337405-25G A337405-100G A337405-500G
103810-166EA 65 USD InStock
103810-166 76839-728 76978-362 77881-930
(2R,3R,4S,5R)-6-Oxohexane-1,2,3,4,5-pentayl pentaacetate 97%
(2R,3R,4S,5R)-6-Oxohexane-1,2,3,4,5-pentayl pentaacetate
Precaución: For research use only


Formula: C₁₆H₂₂O₁₁
Boiling Pt: 434.8 °C (760 mm Hg)
Melting Pt: 109…111 °C
Storage Temperature: Freezer
MDL Number: MFCD09996912
CAS Number: 3891-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty