Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Threoninamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A216728-1G A216728-5G A216728-1G A216728-25G A216728-5G A216728-100G A216728-250MG
103818-580EA 6 USD InStock
103818-580 103849-302 77087-902 77087-904 77087-906 77288-102 77288-104
L-Threoninamide hydrochloride 97%
L-Threoninamide hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₁ClN₂O₂
Boiling Pt: 384.9 °C (760 mm Hg)
Flash Pt: 186.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00058288
CAS Number: 33209-01-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty