Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A101380-1G A101380-5G A101380-100MG A101380-250MG
103795-944EA 55 USD InStock
103795-944 103821-628 77008-320 77008-322
Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid 98%
Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₀H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD02682328
CAS Number: 189249-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty