Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A121153-1G A121153-100MG A121153-250MG A121153-5G A121153-1G
103800-038EA 110 USD InStock
103800-038 77136-490 77136-492 77136-494 77297-818
Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid 95%
Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₆H₂₁NO₄
Boiling Pt: 441.5 °C (760 mm Hg)
Flash Pt: 220.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321010
CAS Number: 96314-29-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty