Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2Z)-4-Oxo-4-[3-(trifluoromethyl)-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-yl]-1-(2,4,5-trifluorophenyl)but-2-en-2-amine 95%

Supplier: AMBEED, INC
Warning

A180769-1G A180769-5G A180769-5G A180769-100G A180769-1G
103807-556EA 14 USD InStock
103807-556 103845-918 77083-420 77138-704 77301-694
(2Z)-4-Oxo-4-[3-(trifluoromethyl)-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-yl]-1-(2,4,5-trifluorophenyl)but-2-en-2-amine 95%
(2Z)-4-Oxo-4-[3-(trifluoromethyl)-5,6-dihydro[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-yl]-1-(2,4,5-trifluorophenyl)but-2-en-2-amine
Precaución: for Research Use Only


Formula: C₁₆H₁₃F₆N₅O
Boiling Pt: 555.1 °C (760 mm Hg)
Melting Pt: 178…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031551
CAS Number: 767340-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty