Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis,cis-Muconic acid, (max. 3% H₂O) 97% (contains < 3% H₂O)

Supplier: AMBEED, INC
Warning

A236064-1G A236064-5G A236064-10G A236064-250MG A236064-25G A236064-1G A236064-5G
103811-066EA 30 USD InStock
103811-066 103850-836 77013-654 77013-656 77013-658 77089-872 77089-874
cis,cis-Muconic acid, (max. 3% H₂O) 97% (contains < 3% H₂O)
cis,cis-Muconic acid
Precaución: for Research Use Only


Formula: C₆H₆O₄
Melting Pt: 194…195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085266
CAS Number: 1119-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty