Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(9H-Carbazol-9-yl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A163213-1G A163213-5G A163213-100G A163213-10G A163213-250MG A163213-1G A163213-25G A163213-5G
103805-698EA 9 USD InStock
103805-698 103843-956 76812-864 76812-866 76812-868 77320-044 77320-046 77320-048
3-(9H-Carbazol-9-yl)phenylboronic acid 97%
3-(9H-Carbazol-9-yl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₄BNO₂
Flash Pt: 239.6 °C
Storage Temperature: Ambient
MDL Number: MFCD18072506
CAS Number: 864377-33-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty