Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 97%

Supplier: AMBEED, INC
Warning

A265095-1G A265095-25G A265095-100G A265095-10G A265095-500G
103812-704EA 7 USD InStock
103812-704 103852-598 77092-262 77092-264 77140-824 77140-826 77140-828
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane 97%
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane
Precaución: for Research Use Only


Formula: C₁₃H₁₃F₁₇O₃Si
Boiling Pt: 247 °C (760 mm Hg)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD07368748
CAS Number: 83048-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty