Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxybenzophenone 98%

Supplier: AMBEED, INC
Warning

A716345-5G A716345-25G A716345-10G
103840-712EA 7 USD InStock
103840-712 76844-926 77170-818
3,4-Dimethoxybenzophenone 98%
3,4-Dimethoxybenzophenone
Precaución: for Research Use Only


Formula: C₁₅H₁₄O₃
Boiling Pt: 380.7 °C (760 mm Hg)
Flash Pt: 175.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00075875
CAS Number: 4038-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty