Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(methoxycarbonyl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A256707-1G A256707-5G A256707-10G A256707-1G A256707-25G A256707-5G
103812-280EA 17 USD InStock
103812-280 103852-144 76874-728 77091-644 77091-646 77091-648
3,5-Bis(methoxycarbonyl)phenylboronic acid 97%
3,5-Bis(methoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BO₆
MW: 238.00 g/mol
Boiling Pt: 436.3 °C (760 mm Hg)
Flash Pt: 217.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11053854
CAS Number: 177735-55-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty