Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-fluorobenzylalkohol 97%

Supplier: AMBEED, INC
Warning

A298917-1G A298917-5G A298917-250MG A298917-1G A298917-5G
103814-202EA 106 USD InStock
103814-202 103835-350 76787-688 77305-754 77305-756
3-Amino-2-fluorobenzylalkohol 97%
3-Amino-2-fluorobenzylalkohol
Precaución: for Research Use Only


Formula: C₇H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD11846488
CAS Number: 1051899-73-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty