Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A772466-5G A772466-10G A772466-1G A772466-25G A772466-5G
103840-942EA 12 USD InStock
103840-942 77118-264 77308-298 77308-300 77308-302
3-Aminophenylboronic acid 98%
3-Aminophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₈BNO₂
Boiling Pt: 376 °C (760 mm Hg)
Melting Pt: 98 °C at N/A mmHg
Flash Pt: 181.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00007755
CAS Number: 30418-59-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty