Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-5-Norbornene exo-2,3-dicarboxylic anhydride 98%

Supplier: AMBEED, INC
Danger

A138981-100G A138981-10G A138981-25G A138981-5G
76688-946EA 21 USD InStock
76688-946 76688-948 77299-038 77299-040
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 98%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride
Precaución: For research use only.


Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 331.1 °C (760 mm Hg)
Melting Pt: 140…145 °C
Flash Pt: 163.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00630580
CAS Number: 2746-19-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty