Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3aS,4S,6aR)-4-Methoxytetrahydrofuro[3,4-b]furan-2(3H)-one 97%

Supplier: AMBEED, INC
Warning

A214271-1G A214271-10G A214271-1G A214271-25G
103818-384EA 9 USD InStock
103818-384 103849-090 76986-852 77087-600 77087-602
(3aS,4S,6aR)-4-Methoxytetrahydrofuro[3,4-b]furan-2(3H)-one 97%
(3aS,4S,6aR)-4-Methoxytetrahydrofuro[3,4-b]furan-2(3H)-one
Precaución: for Research Use Only


Formula: C₇H₁₀O₄
Melting Pt: 126…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698799
CAS Number: 866594-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty