Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluorophenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A228658-1G A228658-5G A228658-250MG A228658-1G A228658-5G
103819-410EA 7 USD InStock
103819-410 103850-314 76815-104 77089-118 77303-832
3-Bromo-2-fluorophenylboronic acid 95%
3-Bromo-2-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BBrFO₂
Boiling Pt: 326.6 °C (760 mm Hg)
Melting Pt: 225…227 °C
Flash Pt: 151.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664228
CAS Number: 352535-97-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty