Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluorobenzylalkohol 98%

Supplier: AMBEED, INC
Warning

A776689-1G A776689-5G A776689-100G A776689-10G A776689-25G
103840-952EA 24 USD InStock
103840-952 76845-208 76845-210 77006-376 77006-378 77308-304
3-Bromo-2-fluorobenzylalkohol 98%
3-Bromo-2-fluorobenzylalkohol
Precaución: for Research Use Only


Formula: C₇H₆BrFO
MW: 205.0200 g/mol
Boiling Pt: 263 °C (760 mm Hg)
Flash Pt: 112.999 °C
Storage Temperature: Ambient
MDL Number: MFCD09842441
CAS Number: 261723-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty