Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromopropoxy)-tert-butyldimethylsilane 97%

Supplier: AMBEED, INC
Warning

A103416-1G A103416-5G A103416-100G A103416-10G A103416-25G A103416-500G A103416-1G A103416-5G
103796-546EA 5 USD InStock
103796-546 103822-252 77135-602 77135-604 77135-606 77135-608 77283-090 77309-120
(3-Bromopropoxy)-tert-butyldimethylsilane 97%
(3-Bromopropoxy)-tert-butyldimethylsilane
Precaución: for Research Use Only


Formula: C₉H₂₁BrOSi
MW: 253.26 g/mol
Boiling Pt: 219.2 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216589
CAS Number: 89031-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty