Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: AMBEED, INC
Warning

A376413-1G A376413-5G A376413-10G A376413-250MG A376413-1G A376413-5G
103826-580EA 23 USD InStock
103826-580 103837-606 77155-262 77155-264 77306-876 77306-878
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)boronic acid 97%
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₄BClF₄O₂
MW: 242.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25459282
CAS Number: 2096342-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty