Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A180558-5G A180558-1G A180558-5G A180558-100G A180558-10G
103807-544EA 12 USD InStock
103807-544 103845-904 77083-398 77083-400 77236-554 77236-556
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid 97%
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BFO₄
Boiling Pt: 195.3 °C (760 mm Hg)
Melting Pt: 180…184 °C
Flash Pt: 92.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04115665
CAS Number: 505083-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty