Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(trifluoromethyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A107949-1G A107949-5G A107949-250MG A107949-25G A107949-1G A107949-5G A107949-100G
103797-272EA 16 USD InStock
103797-272 103823-046 77025-560 77025-562 77283-668 77309-886 77871-423
3-Fluoro-4-(trifluoromethyl)phenylboronic acid 98%
3-Fluoro-4-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₅BF₄O₂
Boiling Pt: 269.8 °C (760 mm Hg)
Flash Pt: 117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08706261
CAS Number: 864759-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty