Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A468483-1G A468483-5G A468483-5G A468483-100G A468483-10G A468483-500G A468483-1G A468483-25G
103828-002EA 7 USD InStock
103828-002 103839-076 76842-982 77116-336 77116-338 77116-340 77307-460 77307-462
3-Hydroxyphenylboronic acid 98%
3-Hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BO₃
Boiling Pt: 370 °C (760 mm Hg)
Melting Pt: 210…213 °C
Flash Pt: 177.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01074603
CAS Number: 87199-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty