Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Methoxybenzyl)triphenylphosphonium chloride 95%

Supplier: AMBEED, INC
Warning

A138751-1G A138751-10G A138751-250MG A138751-1G A138751-25G A138751-5G
103802-660EA 6 USD InStock
103802-660 77272-376 77272-378 77299-014 77299-016 77299-018
(3-Methoxybenzyl)triphenylphosphonium chloride 95%
(3-Methoxybenzyl)triphenylphosphonium chloride
Precaución: for Research Use Only


Formula: C₂₆H₂₄ClOP
MW: 418.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836189
CAS Number: 18880-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty