Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrophenylhydrazinium chloride 95%

Supplier: AMBEED, INC
Danger

A139263-1G A139263-10G A139263-1G A139263-25G A139263-5G
103802-720EA 9 USD InStock
103802-720 77109-746 77299-066 77315-800 77315-802
3-Nitrophenylhydrazinium chloride 95%
3-Nitrophenylhydrazinium chloride
Precaución: for Research Use Only


Formula: C₆H₈ClN₃O₂
Boiling Pt: 317.5 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 145.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00012939
CAS Number: 636-95-3
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty