Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide 98%

Supplier: AMBEED, INC
Warning

A441257-1G A441257-5G A441257-250MG A441257-1G A441257-25G A441257-5G
103827-672EA 18 USD InStock
103827-672 103838-738 76818-248 77307-310 77307-312 77307-314
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide 98%
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₄H₂₆N₂O
Boiling Pt: 401.5 °C (760 mm Hg)
Melting Pt: 112…115 °C
Flash Pt: 146.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01313226
CAS Number: 136465-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty