Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%

Supplier: AMBEED, INC
Warning

A454850-1G A454850-5G A454850-25G A454850-5G A454850-100G A454850-10G
103827-826EA 13 USD InStock
103827-826 103838-896 76842-740 76842-742 76877-550 76877-552
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
Melting Pt: 94…100 °C
Flash Pt: 195.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01073893
CAS Number: 90365-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty