Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Bromomethyl)benzophenone 97%

Supplier: AMBEED, INC
Danger

A201683-1G A201683-5G A201683-10G A201683-25G A201683-1G A201683-5G
103817-316EA 29 USD InStock
103817-316 103847-958 76785-366 77086-034 77302-776 77302-778
4-(Bromomethyl)benzophenone 97%
4-(Bromomethyl)benzophenone
Precaución: for Research Use Only


Formula: C₁₄H₁₁BrO
MW: 275.1405 g/mol
Flash Pt: 79.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00092076
CAS Number: 32752-54-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty