Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Difluoromethoxy)benzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A212028-1G A212028-5G A212028-100MG A212028-10G A212028-250MG A212028-1G A212028-5G A212028-25G
103818-158EA 44 USD InStock
103818-158 103848-854 77030-534 77030-536 77030-538 77087-296 77087-298 77872-081
4-(Difluoromethoxy)benzeneboronic acid 95%
4-(Difluoromethoxy)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09864667
CAS Number: 688810-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty