Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylcarbamoyl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A179852-1G A179852-5G A179852-100G A179852-250MG A179852-1G A179852-5G A179852-25G
103807-482EA 18 USD InStock
103807-482 103845-836 76690-510 76690-512 77083-310 77083-312 77301-654
4-(Dimethylcarbamoyl)phenylboronic acid 97%
4-(Dimethylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BNO₃
Boiling Pt: 407.4 °C (760 mm Hg)
Melting Pt: 130…136 °C
Flash Pt: 200.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02258943
CAS Number: 405520-68-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty