Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methoxycarbonyl)-2-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A148335-1G A148335-100MG A148335-10G A148335-250MG A148335-5G A148335-1G A148335-25G
103803-916EA 24 USD InStock
103803-916 76972-952 76972-954 76972-956 76972-958 77317-482 77881-157
4-(Methoxycarbonyl)-2-methylphenylboronic acid 98%
4-(Methoxycarbonyl)-2-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD16036117
CAS Number: 158429-38-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty