Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4,4-Difluorocyclohexyl)methanol 95%

Supplier: AMBEED, INC
Warning

A198439-1G A198439-5G A198439-10G A198439-250MG A198439-1G A198439-25G A198439-5G
103817-018EA 11 USD InStock
103817-018 103847-644 76825-474 76825-476 77302-584 77302-586 77302-588
(4,4-Difluorocyclohexyl)methanol 95%
(4,4-Difluorocyclohexyl)methanol
Precaución: for Research Use Only


Formula: C₇H₁₂F₂O
Boiling Pt: 181.4 °C (760 mm Hg)
Flash Pt: 63.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847775
CAS Number: 178312-48-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty