Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chlorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A286022-1G A286022-5G A286022-100MG A286022-250MG A286022-5G
103813-700EA 105 USD InStock
103813-700 103834-836 76875-386 76875-388 77305-498
4-Bromo-2-chlorophenylboronic acid 97%
4-Bromo-2-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BBrClO₂
MW: 235.2700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195646
CAS Number: 1046861-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty