Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromo-2-nitrophenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A101956-1G A101956-5G A101956-100G A101956-100MG A101956-10G A101956-250MG A101956-25G A101956-1G A101956-5G
103796-226EA 38 USD InStock
103796-226 103821-920 77233-110 77233-112 77233-114 77233-116 77233-118 77282-872 77282-874
(4-Bromo-2-nitrophenyl)acetic acid 97%
(4-Bromo-2-nitrophenyl)acetic acid
Precaución: for Research Use Only


Formula: C₈H₆BrNO₄
MW: 260.0400 g/mol
Boiling Pt: 392 °C (760 mm Hg)
Melting Pt: 156…160 °C
Flash Pt: 190.9 °C
Storage Temperature: Ambient
MDL Number: MFCD05663950
CAS Number: 6127-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty